Orders LinkContactUsLinkComponent
Visual comparator colour kits, phenols
Visual comparator colour kits, phenols
Phenols test kit, model PL-1, 0 - 4 mg/l, 4-Aminoantipyrine, 1 KIT
Catalogus # HACH2483600
Leverancier:   Hach
Visual comparator colour kits, phenols
Catalogus # HACH2483600
Leverancier:  Hach
CAS-nummer:  
Visual comparator colour kits, phenols
Visual comparator colour kits, phenols
Phenols test kit, model PL-1, 0 - 4 mg/l, 4-Aminoantipyrine, 1 KIT
CAS-nummer:  
Price
312,00 €
Log in voor beschikbaarheid
1 * 1 KIT
Product Details & Documents
Simple, economical, rapid visual colour determination.
 : Kit includes four reagents, two tubes, two color discs, color comparator box, instruction sheet, and carrying case.
Specificaties
Specificaties
  • Chemische methode:
    4-Aminoantipyrine
  • VPE:
    1 KIT
  • Omschrijving:
    Phenols test kit, model PL-1
  • Meetbereik:
    0 - 4 mg/l
Specificaties