Caps for culture tubes
Catalog # 705-1201
Supplier: Hach
Specifications
- Schroefdraad:15-415
- Kleur:wit
- Omschrijving:Screw cap PP with fluoropolymer liner
- Environmentally Preferable:
- VPE:6
Specifications
About this item
Screw cap with fluoropolymer liner.
- Firmly bonded liner provides a highly inert sealing surface