Orders LinkContactUsLinkComponent
m-HPC broth
m-HPC broth
m-HPC broth, plastic ampules, Total Count, 50
Catalogus # HACH2812450
Leverancier:   Hach
m-HPC broth
Catalogus # HACH2812450
Leverancier:  Hach
CAS-nummer:  
m-HPC broth
m-HPC broth
m-HPC broth, plastic ampules, Total Count, 50
CAS-nummer:  
Price
216,00 €
Log in voor beschikbaarheid
1 * 50 ST
Product Details & Documents
The m-HPC broth used in heterotrophic plate count (HPC), for estimating the number of live heterotrophic bacteria in water. This test can provide useful information about water quality and supporting data on the significance of coliform test results. High concentrations of the general bacterial population may hinder the recovery of coliforms
  • Used to judge the efficiency of various treatment processes for both drinking water and swimming pools
  • Checking the quality of finished water in a distribution system
  • Save time and money by using prepared media for membrane filtration methods
Specificaties
Specificaties
  • VPE:
    50
  • Omschrijving:
    m-HPC broth, plastic ampules
  • Voor:
    Total Count