Orders LinkContactUsLinkComponent
Molybdate test kit, MO-LR
Molybdate test kit, MO-LR
Molybdate test kit, MO-LR, 0 - 3,0 mg/l Mo, 1 KIT
Catalogus # HACH2359300
Leverancier:   Hach
Molybdate test kit, MO-LR
Catalogus # HACH2359300
Leverancier:  Hach
CAS-nummer:  
Molybdate test kit, MO-LR
Molybdate test kit, MO-LR
Molybdate test kit, MO-LR, 0 - 3,0 mg/l Mo, 1 KIT
CAS-nummer:  
Price
311,00 €
Log in voor beschikbaarheid
1 * 1 KIT
Product Details & Documents
Molybdates are used to inhibit corrosion in industrial and utility water systems. The reagents in the kit reacts with molybdate to form a blue colour, which is measured with a colour disc.
  • Easy to use colour disc
  • Portable test kit for on-site measurement
  • Effectively monitors the levels of molybdate corrosion inhibitors
Levering: The kit includes two reagents, one color comparator box, one color disc, two stoppers, instructions, and a carrying case.
Specificaties
Specificaties
  • VPE:
    1 KIT
  • Omschrijving:
    Molybdate test kit, MO-LR
  • Meetbereik:
    0 - 3,0 mg/l Mo
Specificaties