Lysates, human
Over dit artikel
Whole cell lysates are derived from cell lines tissues using highly refined extraction protocols to ensure quality, protein integrity and high reproducibility. Lysates are prepared in denaturing buffer without dissociating agents.
- Liquid
Whole cell lysates are ready to use for SDS-PAGE and Western blotting purposes. All extracts are tested by SDS-PAGE using 4 to 20% gradient gels and immunoblot analysis using antibodies to key cell signaling components to confirm the presence of both high molecular weight and low molecular weight proteins.
Each vial contains 1mg/ml of protein in SDS-PAGE sample buffer. The sample buffer consists of 0,05 mM Tris Hcl, 2% SDS, 10% glycerol and 0,1% bromophenol blue at pH 6,8.
2 Opties Hieronder Beschikbaar
Product Details & Documents
Whole cell lysates are derived from cell lines tissues using highly refined extraction protocols to ensure quality, protein integrity and high reproducibility. Lysates are prepared in denaturing buffer without dissociating agents.
- Liquid
Whole cell lysates are ready to use for SDS-PAGE and Western blotting purposes. All extracts are tested by SDS-PAGE using 4 to 20% gradient gels and immunoblot analysis using antibodies to key cell signaling components to confirm the presence of both high molecular weight and low molecular weight proteins.
Each vial contains 1mg/ml of protein in SDS-PAGE sample buffer. The sample buffer consists of 0,05 mM Tris Hcl, 2% SDS, 10% glycerol and 0,1% bromophenol blue at pH 6,8.
Documents
Product Family Options
Productinformatie
- VPEBeschikbaarheidPrice
- ROCKW09-000-360MCF-7 whole cell lysateMCF-7 whole cell lysate500 µGDiscontinued-
- ROCKW09-000-366MCF-7 whole cell lysate, hydrogen peroxide stimulatedMCF-7 whole cell lysate, hydrogen peroxide stimulated500 µGDiscontinued-
- ROCKW09-000-361A431 whole cell lysateA431 whole cell lysate500 µGDiscontinued-
- ROCKW09-000-362A431 whole cell lysate - EGF stimulatedA431 whole cell lysate - EGF stimulated500 µGDiscontinued-
- ROCKW09-000-363Hep2 whole cell lysateHep2 whole cell lysate500 µGDiscontinued-
- ROCKW09-000-364HeLa whole cell lysateHeLa whole cell lysate500 µGDiscontinued-
- ROCKW09-000-365HEK293, whole cell lysateHEK293, whole cell lysate500 µGDiscontinued-
- ROCKW09-001-370Jurkat whole cell lysate
- ROCKW09-001-GY4U251 Whole Cell Lysate