Anti-vibration pad, Richter
Catalog # DIVEVIBE-1000
Supplier: DIVERSIFIED BIOTECH
Specifications
- Omschrijving:Anti-vibration pad
- l×b:215x170 mm mm
- VPE:1
Specifications
About this item
Richter anti-vibration pad dampens vibrations generated from lab equipment. It creates a stable surface that will keep equipment in place.
- Small, lightweight design makes transportation between benches and laboratories easy
- Pads can be stacked for heavier units or laid side-by-side for use with larger equipment
Ideal for use with vortexes, shakers, rockers and microscopes.