Desktop stapler
Desktop stapler, zwart, Metaal
Catalogus # 818-3141
Leverancier: WEDO WERNER DORSCH
Ga naar:
Product Details & Documents
Plastic grip stapler, specially designed with side-hinged stapler arm for stapling booklets, brochures, leaflets, etc.
- Capacity of 25 sheets (80 gsm)
- Dimensions: 147×40×60 mm
- Weight: 210 g
- Staple size 24/6, 26/6
Specificaties
- Kleur:zwart
- Omschrijving:Desktop stapler
- Materiaal:Metaal