Safety cap
Safety cap, with PTFE insert and EPDM O-ring, GL 45, 2× 1/8" tubing port, 1× 1/16" tubing port, 1× 1/4"-28 female port, PP
Catalogus # JOURJR-9000-0001
Leverancier: VICI JOUR RESEARCH
Ga naar:
Product Details & Documents
This cap can be used for helium sparging and for delivering HPLC mobile phases. The tubing ports are made so that you push the tubing through the hole, while 1/4-28 fittings provide the best connection. Unused ports can be plugged as required.
Specificaties
- Schroefdraad:GL 45
- Omschrijving:Safety cap, with PTFE insert and EPDM O-ring
- Materiaal:PP
- Seq:01
- Slangverbinding:2× 1/8" tubing port, 1× 1/16" tubing port, 1× 1/4"-28 female port