Orders LinkContactUsLinkComponent
Fraction-FOCUS™
Fraction-FOCUS™
Fraction-FOCUS™, 10 Tests
Catalogus # 786-168
Leverancier:   G-Biosciences
Fraction-FOCUS™
Catalogus # 786-168
Leverancier:  G-Biosciences
CAS-nummer:  
Fraction-FOCUS™
Fraction-FOCUS™
Fraction-FOCUS™, 10 Tests
CAS-nummer:  
Price
736,00 €
Log in voor beschikbaarheid
1 * 1 EA
Product Details & Documents
Fraction-FOCUS™ utilises proven technology to fractionate and concentrate all proteomes into multiple fractions; simplifying 2D maps and enhancing detection of low abundant proteins. Fraction-FOCUS™ is fully compatible with all downstream protein identification techniques.
There is no detectable loss of total protein during the Fraction-FOCUS™ procedure. At the end of the fractionation, cellular proteins are in one of many fractions.
Documents
Specificaties
Specificaties
  • VPE:
    10 Tests
  • Omschrijving:
    Fraction-FOCUS™
Specificaties
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.