Orders LinkContactUsLinkComponent
Foot adapter
Foot adapter
Foot adapter, aluminium, 13 mm
Catalogus # RLAA2119
Leverancier:   R L ENTERPRISES
Foot adapter
Catalogus # RLAA2119
Leverancier:  R L ENTERPRISES
Foot adapter
Foot adapter, aluminium, 13 mm
Price
31,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Adapter manufactured out of aluminium with boss to fit scaffold feet.
Accepts three 13 mm diameter rods or tubes at right angles (2 perpendicular to the boss and the third in line with the boss), with alien screws for fixing. One allen key provided per pack of ten.
Specificaties
  • Omschrijving:
    Foot adapter, aluminium
  • Voor staaf Ø:
    13 mm