Surface cleaner, Mr Proper, P&G Professional
Catalogus # KING050208/00
Leverancier: KING DISPOSABLES
Specificaties
- Omschrijving:Mr Proper, Ocean, surface cleaner
- Environmentally Preferable:
- VPE:5 L
Specificaties
Over dit artikel
Liquid concentrate for cleaning all washable hard surface, especially floors.
- Less than 5% of non-ionic surfactants, soap, glutaraldehyde, benzisothiazolinone
- Fresh residual fragrance
- Effective action on grease, and dirt particles, regardless of the type of water
- Rinsing is unnecessary when the product is diluted