Orders LinkContactUsLinkComponent
Precision ground-joint thermometers
Precision ground-joint thermometers
Leverancier:   Ludwig Schneider
Precision ground-joint thermometers
Leverancier:  Ludwig Schneider
Certificaten
Precision ground-joint thermometers
Precision ground-joint thermometers
25 Opties Hieronder Beschikbaar
Product Details & Documents
These precision ground-joint thermometers are suitable for official certification.
  • Colourless, prism capillary form
  • Upper part: Ø 11 ±0,5 mm, with ground joint NS 14,5/23
Blue, red, pentane, alcohol, mercury or toluene filling, enclosed type.
Conforms to DIN 12784.
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.
Certifications: Conforms to DIN 12784.
Ordering information: Available with works certificate or with DAkkS calibration certificate (optional).
Packaging: Single packed in plastic square outer covering.
Services: Voor installatie, IQOQ, erkende kalibratie onder ISO17025, onderhoud en reparatie mail naar [email protected]
Product Family Options
Blue filling
Productinformatie
  • Meetbereik
    Indeling
    Lengte
    Vulling
    Onderdompeldiepte
    Voor NS
    Beschikbaarheid
    Price
  • SCNL1292611
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −38...+50 °C
    1,0 °C
    325 mm
    Blue
    52 mm
    14,5/23
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 58,10 €
    58,10 €
    1 * 1 ST
       58,10 €
  • SCNL1292612
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −38...+50 °C
    1,0 °C
    345 mm
    Blue
    72 mm
    14,5/23
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 58,60 €
    58,60 €
    1 * 1 ST
       58,60 €
  • SCNL1292613
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −38...+50 °C
    1,0 °C
    375 mm
    Blue
    102 mm
    14,5/23
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 63,50 €
    63,50 €
    1 * 1 ST
       63,50 €
  • SCNL1292614
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −38...+50 °C
    1,0 °C
    400 mm
    Blue
    127 mm
    14,5/23
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 63,70 €
    63,70 €
    1 * 1 ST
       63,70 €
  • SCNL1292615
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −38...+50 °C
    1,0 °C
    425 mm
    Blue
    152 mm
    14,5/23
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 65,10 €
    65,10 €
    1 * 1 ST
       65,10 €
  • SCNL1292616
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −38...+50 °C
    1,0 °C
    450 mm
    Blue
    177 mm
    14,5/23
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 65,70 €
    65,70 €
    1 * 1 ST
       65,70 €
  • SCNL1302210
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −10...+150 °C
    1,0 °C
    318 mm
    Blue
    45 mm
    14,5/23
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 55,20 €
    55,20 €
    1 * 1 ST
       55,20 €
  • SCNL1302211
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −10...+150 °C
    1,0 °C
    325 mm
    Blue
    52 mm
    14,5/23
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 55,50 €
    55,50 €
    1 * 1 ST
       55,50 €
  • SCNL1302212
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −10...+150 °C
    1,0 °C
    345 mm
    Blue
    72 mm
    14,5/23
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 55,80 €
    55,80 €
    1 * 1 ST
       55,80 €
  • SCNL1302213
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −10...+150 °C
    1,0 °C
    375 mm
    Blue
    102 mm
    14,5/23
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 60,80 €
    60,80 €
    1 * 1 ST
       60,80 €
  • SCNL1302214
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −10...+150 °C
    1,0 °C
    400 mm
    Blue
    127 mm
    14,5/23
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 61,10 €
    61,10 €
    1 * 1 ST
       61,10 €
  • SCNL1302215
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −10...+150 °C
    1,0 °C
    425 mm
    Blue
    152 mm
    14,5/23
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 62,20 €
    62,20 €
    1 * 1 ST
       62,20 €
  • SCNL1302216
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −10...+150 °C
    1,0 °C
    450 mm
    Blue
    177 mm
    14,5/23
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 62,90 €
    62,90 €
    1 * 1 ST
       62,90 €
  • SCNL1302410
    Enclosed type, with colourless prismatic capillary form, without official certification
    Enclosed type, with colourless prismatic capillary form, without official certification
    −10...+250 °C
    1,0 °C
    368 mm
    Blue
    45 mm
    14,5/23
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 69,40 €
    69,40 €
    1 * 1 ST
       69,40 €
  • SCNL1302411
    Enclosed type, with colourless prismatic capillary form, without official certification
    Enclosed type, with colourless prismatic capillary form, without official certification
    −10...+250 °C
    1,0 °C
    375 mm
    Blue
    52 mm
    14,5/23
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 69,70 €
    69,70 €
    1 * 1 ST
       69,70 €
  • SCNL1302412
    Enclosed type, with colourless prismatic capillary form, without official certification
    Enclosed type, with colourless prismatic capillary form, without official certification
    −10...+250 °C
    1,0 °C
    395 mm
    Blue
    72 mm
    14,5/23
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 70,00 €
    70,00 €
    1 * 1 ST
       70,00 €
  • SCNL1302413
    Enclosed type, with colourless prismatic capillary form, without official certification
    Enclosed type, with colourless prismatic capillary form, without official certification
    −10...+250 °C
    1,0 °C
    425 mm
    Blue
    102 mm
    14,5/23
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 75,00 €
    75,00 €
    1 * 1 ST
       75,00 €
  • SCNL1302414
    Enclosed type, with colourless prismatic capillary form, without official certification
    Enclosed type, with colourless prismatic capillary form, without official certification
    −10...+250 °C
    1,0 °C
    450 mm
    Blue
    127 mm
    14,5/23
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 75,30 €
    75,30 €
    1 * 1 ST
       75,30 €
  • SCNL1302415
    Enclosed type, with colourless prismatic capillary form, without official certification
    Enclosed type, with colourless prismatic capillary form, without official certification
    −10...+250 °C
    1,0 °C
    475 mm
    Blue
    152 mm
    14,5/23
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 76,70 €
    76,70 €
    1 * 1 ST
       76,70 €
  • SCNL1302416
    Enclosed type, with colourless prismatic capillary form, without official certification
    Enclosed type, with colourless prismatic capillary form, without official certification
    −10...+250 °C
    1,0 °C
    500 mm
    Blue
    177 mm
    14,5/23
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 77,30 €
    77,30 €
    1 * 1 ST
       77,30 €
Red filling
Productinformatie
  • Meetbereik
    Indeling
    Lengte
    Vulling
    Onderdompeldiepte
    Voor NS
    Beschikbaarheid
    Price
  • SCNL1292201
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −100...+30 °C
    1,0 °C
    318 mm
    Red
    45 mm
    14,5/23
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 81,00 €
    81,00 €
    1 * 1 ST
       81,00 €
  • SCNL1292202
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −100...+30 °C
    1,0 °C
    325 mm
    Red
    52 mm
    14,5/23
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 81,20 €
    81,20 €
    1 * 1 ST
       81,20 €
  • SCNL1292203
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −100...+30 °C
    1,0 °C
    345 mm
    Red
    72 mm
    14,5/23
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 81,50 €
    81,50 €
    1 * 1 ST
       81,50 €
  • SCNL1292204
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −100...+30 °C
    1,0 °C
    375 mm
    Red
    102 mm
    14,5/23
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 86,60 €
    86,60 €
    1 * 1 ST
       86,60 €
  • SCNL1292304
    Enclosed type, with colourless prismatic capillary form
    Enclosed type, with colourless prismatic capillary form
    −100...+30 °C
    0,5 °C
    375 mm
    Red
    102 mm
    14,5/23
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 104,00 €
    104,00 €
    1 * 1 ST
       104,00 €