Orders LinkContactUsLinkComponent
Cryoboxes
without grid insert, -, tissue culture tubes 15/50 ml, 122×122×128 mm
  NEOL2-7094
 :   NEOLAB
Cryoboxes
  NEOL2-7094
 :  NEOLAB
Cryoboxes
Cryoboxes
without grid insert, -, tissue culture tubes 15/50 ml, 122×122×128 mm
15,12 €
1 * 1 ST
Cryo resistant cardboard.
  • Water repellent
  • Llid has a marking area on one of the sides
For storage of tissue culture tubes of 15 and 50 ml capacities for cryogenic deep freezers.
  • Compartimenten:
    -
  • Voor:
    tissue culture tubes 15/50 ml
  • Uitw. b×d×h:
    122×122×128 mm
  • Omschrijving:
    without grid insert
  • Seq:
    01