Handheld digital microscope, Dino-Lite Medical, IriScope
Catalogus # 630-2615
Leverancier: Dino-Lite / IDCP B.V.
Specificaties
- Vergroting:10 - 20×
- Model:IriScope
- Resolutie:1,3 MP
- Omschrijving:Handheld microscope for iris examinations
- Seq:01
- Verlichting:8 LEDs (white + yellow)
- VPE:1
Specificaties
Over dit artikel
The IriScope enables users to make razor-sharp images of the iris.
- Handy, easy to use
- Magnification of 10 to 20 times allows users to see details that can hardly be seen by the naked eye
- Specially designed front cap can be placed on the eye socket to get the right distance for a perfect picture
- Built-in LEDs (white + yellow) provide better images of dark irises
- High speed USB 2.0 output
Certificeringen: Medical Device Class 1 – according to Directive 93/42/EEC modified 2007/47/EC.
Levering: Supplied with protecting pouch, software CD and calibration target.