Counting machine
Counting machine, 5-place
Catalogus # 720-2761
Leverancier: HAWKSLEY AND SONS
Ga naar:
Product Details & Documents
Totaliser five place counting machine. An intuitive differential cell counter that simplifies blood cell counts.
- When a total of 100 cells is counted, a bell rings
- Percentage of each type of cell can then be read directly from the window above each key
- simple counter-clockwise rotation of the end knobs will zero all the counts
- Each key has the name and picture of a particular cell above it, representing groups of leucocytes
- The key legend strips are easily removed to allow customisation
- Five place set to count Basophils, Eosinophils, Seg., Lymphocytes, Monocytes
- Sturdy solid construction with firm dependable keys
Specificaties
- Omschrijving:Counting machine, 5-place
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.