Silver electrode, ScienceLine
Metal electrode, Ag 1100, Ag 1100, −5...+100 °C, Glas, 4×120 mm, Plug head
Catalog # SCOI285103607
Supplier: SI Analytics
Jump to:
Product Details & Documents
Silver electrode with glass shaft.
Specifications
- Aansluiting:Plug head
- Ø×L:4×120 mm
- Type:Ag 1100
- Omschrijving:Metal electrode, Ag 1100
- Materiaal:Glas
- Temperatuur:−5...+100 °C