Transport and Storage Container
Catalog # SEMA3665
Supplier: SEMADENI
Specifications
- Uitw. b×d×h:340×235×160 mm
- Capaciteit:10 l
- Omschrijving:Transport and storage container
- VPE:1
Specifications
About this item
HDPE, blue.
- Light, robust
- Long lasting
- Stackable for space-saving storage
Multipurpose boxes for a wide range of uses.