ContactUsLinkComponent
Laboratory Containers for Liquids
Laboratory Containers for Liquids
  SEMA2099
 :  SEMADENI
undefined
Laboratory Containers for Liquids
  SEMA2099
 :  SEMADENI
  • Seq:
    01
  • b×d×h:
    220×120×120 mm
  • VPE:
    1
PP, beige.
  • Excellent chemical resistance
  • Temperature resistance up to 90 °C
  • Strong-walled
Ideal for use as chemical or water bath.