Laboratory reactor system
Laboratory reactor system, 220 °C, 6 bar, complete, includes 2 l glass vessel, 316 stainless steel lid fitted with stirrer head and heating jacket, pressure gauge, excess pressure valve, thermometer pocket, B29 adaptor with pressure tight cap, inlet with plug valve, tripod and a safety cage
Catalogus # JUCH10101.12
Leverancier: JUCHHEIM Laborgerate GmbH
Ga naar:
Product Details & Documents
Specificaties
- Omschrijving:Laboratory reactor system, 220 °C, 6 bar, complete, includes 2 l glass vessel, 316 stainless steel lid fitted with stirrer head and heating jacket, pressure gauge, excess pressure valve, thermometer pocket, B29 adaptor with pressure tight cap, inlet with plug valve, tripod and a safety cage
Accessories
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Lid, stainless steel, without fittingsLid, stainless steel, without fittingsJUCH10101.20Log in voor beschikbaarheid1 * 1 ST - 6 240,00 €
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Spare bolt springs, each
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Spare compression ring
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Spare ‘O’ ring
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Spare pressure equalising spring
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Rear wall, additional, wire netting
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Safety cage, three sided
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Glass vessel body with jacket, 2 l
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Gaskets, PTFE, for conical joint adaptors B29 with caps, Ø 36×30 mmGaskets, PTFE, for conical joint adaptors B29 with caps, Ø 36×30 mmJUCH20336.31Log in voor beschikbaarheid1 * 1 ST - 13,30 €
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Double joint, stainless steel