Spatulas, disposable
Spatula, short, with broad ending, 115 mm, 22 mm, wit
Catalogus # SEMA3339
Leverancier: SEMADENI
Ga naar:
Product Details & Documents
PS, white. These disposable spatulas are ideal for handling creams, paints or other viscous media.
- Durable, slightly elastic
- Available in different sizes and designs
Specificaties
- Kleur:wit
- Omschrijving:Spatula, short, with broad ending
- Lengte:115 mm
- Seq:02
- Breedte blad:22 mm