Prefilter attachment
Prefilter attachment, Roestvrij staal
Catalogus # SART16807
Leverancier: Sartorius
Ga naar:
Product Details & Documents
Allows the separation of coarse solids from samples with bacteriological germ detection in a single operation, before and during the actual germ-retaining filtration step.
Specificaties
- Omschrijving:Prefilter attachment
- Materiaal:Roestvrij staal
- Seq:01