Jars with overlapping lid
46 mm, 42 mm
Catalogus # SEMA2055
Leverancier: SEMADENI
Ga naar:
Product Details & Documents
Jars, made of PS, clear, with closing lid.
- Good chemical resistance
- Easy to clean
Suitable for ointments, powders, and granules.
Specificaties
- Ø uitw.:46 mm
- Hoogte:42 mm
- Seq:12