Orders LinkContactUsLinkComponent
Erlenmeyer flasks, wide neck
Erlenmeyer flasks, wide neck
Supplier:   SMITH SCIENTIFIC
Erlenmeyer flasks, wide neck
Supplier:  SMITH SCIENTIFIC
Certificates
Erlenmeyer flasks, wide neck
2 Options Available Below
Product Details & Documents
Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With beaded rim and graduation.
Product Family Options
Product Information
  • Capaciteit
    Hoogte
    Ø uitw.
    Ø uitw. hals
    Availability
    Price
  • 250 ml
    140 mm
    85 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 111,00 €
    111,00 €
    1 * 10 ST
       111,00 €
  • 500 ml
    175 mm
    105 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,00 €
    16,00 €
    1 * 1 ST
       16,00 €