Orders LinkContactUsLinkComponent
Pocket thermometer, QUICKFIT®
Pocket thermometer, QUICKFIT®
Pocket thermometer for Quickfit® still heads, 14/23 cone size, 45 mm
Catalogus # 620-2286
Leverancier:   SciLabware
Pocket thermometer, QUICKFIT®
Catalogus # 620-2286
Leverancier:  SciLabware
Pocket thermometer, QUICKFIT®
Pocket thermometer, QUICKFIT®
Pocket thermometer for Quickfit® still heads, 14/23 cone size, 45 mm
Price
32,90 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Quickfit® ground-glass joints are precision manufactured with an accurate taper and smooth finish for leak-free performance and assured interchangeability.
  • For incorporating thermometers into Quickfit® jointed glassware assemblies
  • Pocket fits most Quickfit® still heads
Specificaties
  • Omschrijving:
    Pocket thermometer for Quickfit® still heads, 14/23 cone size, 45 mm