Gas cylinder holder, Sekuroka®
ROTH1513.1
- Type:Gas cylinder support
- VPE:1
This gas cylinder holder is suitable for safe attachment of gas bottles up to 350 mm Ø. Can be easily fastened onto workbenches, tables, shelves or a ledge up to 60 mm thickness.
- Bore hole for bolt
- Variable attachment with black nylon strap