Digital refractometers
0 − 45 °Bx, ±0,001 °Bx
Catalogus # 635-0068
Leverancier: EUROMEX
Ga naar:
Product Details & Documents
Digital refractometer with automatic temperature compensation (ATC) for sugar concentration.
Levering: Supplied with a nine-volt battery.
Specificaties
- Meetbereik:0 − 45 °Bx
- Precisie:±0,001 °Bx