Orders LinkContactUsLinkComponent
Test gas cans
 :   M S A
Test gas cans
 :  M S A
Test gas cans
Test gas cans
These test gases are suitable for use with wearable gas devices.
Product Family Options
  • Omschrijving
  • Test gas, 50 ppm NH3 in N2, capacity 34 l
    1 * 1 ST
    1 * 1 ST
  • Test gas, CH4 - 1.45%, CO2 - 2.5%, CO - 60, H2S - 20, O2 - 15%, capacity 58 l