Safety spectacles dispenser
Safety glasses dispenser, stainless steel, 300×200×400 mm
Catalogus # 113-8174
Leverancier: Palbam
Ga naar:
Product Details & Documents
This dispenser is suitable for use with safety spectacles in the cleanroom industries, such as pharmaceutical, biotechnology, medical device, microelectronics and nanotechnology.
- High-quality design
- Four holes for stable wall mounting
- Clear front panel for higher visibility
- Protection against splashes and impacts
- Stainless steel 304, electropolished finish
Certifications: ISO certified
Delivery information: Typically available from stock for direct shipment from the supplier.
Documents
Palbam Class - Catalog 2018
Product Literature
Specificaties
- Omschrijving:Safety glasses dispenser, stainless steel
- b×d×h:300×200×400 mm
- Cat. No.:113-8174
- Soort dispenser:Wandophanging
- Materiaal:Roestvrij staal
- PPE type:Brillen