Cutter for stainless steel tubing, cutting wheel type
554-1103
- Omschrijving:Tubing cutter, for cutting 1/16" and 1/8" stainless steel tubing, cutting wheel type
- Seq:01
- VPE:1
This is the ideal tool for cutting ¹/₁₆ and ¹/₈" stainless steel tubing, leaving smooth, uniform cuts requiring a minimum of deburring and/or reaming.
The easily replaced cutting wheel scores tubing, leaving cuts which allow tube-to-tube and tube-to-frit connections.