Soxhlet extractor, multiposition
Soxhlet extraction, 100 ml, 6 sample pieces, SMX 100, 100 ml
Catalogus # BEHRB00218423
Leverancier: BEHR
Ga naar:
Product Details & Documents
Complete workstation to undertake Soxhlet extraction using 6×100 ml flasks.
Levering: Workstation comprises temperature and time controller, heating block with six sample positions, condenser frame with coolant water distribution manifold, glassware including flasks, extractors and Dimroth condensers.
Specificaties
- Type:SMX 100
- Omschrijving:Soxhlet extraction, 100 ml, 6 sample pieces
- Kolfinhoud:100 ml