0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Dozen
Dozen voor vials
Storage Boxes for 50 Vials
Storage Boxes for 50 Vials
Catalog # HEWL5182-0575
Supplier:
Agilent
Expand image
Storage Boxes for 50 Vials
Catalog # HEWL5182-0575
Supplier:
Agilent
Specifications
Omschrijving:
Storage boxes for vials, holds 50 vials
VPE:
1
Specifications
About this item
Product Details
Product Details