Hydrometers
Leverancier: NEUBERT VOLUME GLASSWAERE
Ga naar:
Over dit artikel
Both hydrometers (aerometers) are designed for the determination of density by the Archimede's principle.
Certificeringen: DIN 12791
2 Opties Hieronder Beschikbaar
Product Details & Documents
Both hydrometers (aerometers) are designed for the determination of density by the Archimede's principle.
Certificeringen: DIN 12791
Product Family Options
Productinformatie
- MeetbereikIndelingLengteBeschikbaarheidPrice
- NEUB3910-08Hydrometer
- NEUB3910-09Hydrometer