Capillary column end caps
Catalogus # 555-9468
Leverancier: Thermo Fisher Scientific
Specificaties
- Omschrijving:Capillary column end caps, paired
- Seq:01
- VPE:10
Specificaties
Over dit artikel
These capillary column end caps are universal and fit any capillary column.
- Reusable
- Eliminate column contamination caused by leaving a column unsealed or sealed with a septum
- Colour coded to ensure that the column is reinstalled the same way it came out