ContactUsLinkComponent
Accessories for scroll pumps, scrollvac, ILMVAC
Accessories for scroll pumps, scrollvac, ILMVAC
Catalogus # ILMV700145
Accessories for scroll pumps, scrollvac, ILMVAC
Catalogus # ILMV700145
Specificaties
  • Omschrijving:
    Condensate separator, PT 25, to protect scroll pumps from condensable vapours
  • VPE:
    1
Specificaties
Over dit artikel