Bosshead, with burette and tube clamp
Catalogus # 241-0009
Leverancier: SELECTA
Specificaties
- VPE:1
- Voor staaf Ø:7 - 25 mm
Specificaties
Over dit artikel
Bosshead with burette and tube clamp, made of chrome plated metal.
- With PVC coated double prong jaws
- For Ø from 7 to 25 mm