Immersion water heater
Catalog # BREU31430
Supplier: BREUKHOVEN
Specifications
- Verwarmingsvermogen:300 W
- Omschrijving:Immersion heater, length 115 mm, coil Ø×thickness: 37×6 mm, 220 V
- VPE:1
Specifications
About this item
Immersion heater for quickly heating small quantities of water.
- Ideal for situations where open flames should be avoided