Spray bottles
1000 ml
Catalogus # HECH49119010
Leverancier: GLASWARENFABRIK KARL HECHT
Ga naar:
Product Details & Documents
PE spray bottles with adjustable jet.
- Translucent
- Good chemical resistance
- Good temperature resistance
Specificaties
- Capaciteit:1000 ml