Urine collection containers, with self-adhesive lid
Catalogus # BOET06.073.1400
Leverancier: BOETTGER PAUL
Specificaties
- Capaciteit:100 ml
- Omschrijving:Urine cup with printed self-adhesive lid, PP
- VPE:1.000
Specificaties
Over dit artikel
PP, translucent.
- Resistant to pressure and shatterproof, graduated
- Tight sealing lid, no leakage, no inconvenient smells
- Lids can be easily and clearly written on
- Content can be poured out easily after perforation
Ideal for collection and storage of urine samples.
Let op: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904