Patent blue V sodium salt
Catalogus # TCIAA1242-5G
Leverancier: TCI
CAS-nummer:
Specificaties
- Verpakking:Glass bottle
- VPE:5 g
- Seq:3173
Specificaties
Specificatie testresultaten
- Appearance:Red to Dark blue to Black powder to crystal
- Lambda max.:635.0 to 639.0 nm (H2O)
- Absorbance (E1%1cm):min. 1,500 (H2O, 635.0 to 639.0 nm)
Specificatie testresultaten
Over dit artikel
:
:582.67
EINECS:243-654-4
CAS-nummer:20262-76-4
Formule:C₂₇H₃₁N₂NaO₇S₂
MDL-nummer:MFCD00012119