Orders LinkContactUsLinkComponent
Tungsten dichloride dioxide ≥99%
Tungsten dichloride dioxide ≥99%
Certificates
Tungsten dichloride dioxide ≥99%
Tungsten dichloride dioxide ≥99%
2 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 13520-76-8 Molecular Formula: WO2Cl2 Formula Weight: 286.75 Physical Form: Powder Sensitivity: Moisture Sensitive UN No.: 3260 MDL No.: MFCD00054136
  • CAS Number:13520-76-8
  • Formula:Cl₂O₂W
  • MDL Number:MFCD00054136
  • Molecular Weight:286.75 g/mol
  • Storage Temperature:Kamertemperatuur
  • EINECS:236-862-1
  • ADR:8,III
Product Family Options
Product Information