Orders LinkContactUsLinkComponent
Saffron, Q Path®
Saffron, Q Path®
Plastic bottle, 10 g
Catalogus # 11507736.
Leverancier:   VWR Chemicals
Saffron, Q Path®
Catalogus # 11507736.
Leverancier:  VWR Chemicals
CAS-nummer:  84604-17-1
Saffron, Q Path®
Saffron, Q Path®
Plastic bottle, 10 g
CAS-nummer:  84604-17-1
Price
73,30 €
Log in voor beschikbaarheid
1 * 10 g
Product Details & Documents
Dye which can be used for HES histological staining. This product is not IVD registered.
Synoniemen:  C.I. 75100
  • CAS-nummer:84604-17-1
  • Molecular Weight:976.96 g/mol
  • EINECS:283-295-0
Specificaties
Specificaties
  • Verpakking:
    Plastic bottle
  • VPE:
    10 g