ContactUsLinkComponent
Fenolftaleïne 1% in ethanol 90%
Fenolftaleïne 1% in ethanol 90%
Catalog # ROTHK034.3
Supplier:  Roth Carl
CAS Number:  
undefined
Fenolftaleïne 1% in ethanol 90%
Catalog # ROTHK034.3
Supplier:  Roth Carl
CAS Number:  
Specifications
  • Verpakking:
    Glass bottle
  • VPE:
    1 L
  • Seq:
    5037
Specifications
About this item
 :
 :318.33 g/mol
Boiling Pt:∼78 °C (1013 hPa)
Density:∼0.85 g/cm³ (20 °C)
Storage Temperature:Kamertemperatuur
EINECS:201-004-7
UN:1170
ADR:3,II
 :GHS02
 :GHS08
CAS Number:77-09-8
Formula:C₂₀H₁₄O₄
MDL Number:MFCD00005913