Chloramine-T (sodium salt) trihydrate
Catalog # PRLS88873.360
Supplier: PROLABO CMP
CAS Number:
Specifications
- VPE:5 L
- Seq:5190
Specifications
About this item
:
Merck Index:13,02085
Melting Pt:167…170 °C
Density:1.43 g/cm³ (20 °C)
Flash Pt:192 °C (closed cup)
Storage Temperature:Kamertemperatuur
EINECS:204-854-7
UN:3263
ADR:8,III
:281.69 g/mol
:GHS05
:GHS08
:GHS07
CAS Number:7080-50-4
Formula:CH₃C₆H₄SO₂NClNa·3H₂O
MDL Number:MFCD00149066