Microorganism tests, BART test for sulfate-reducing bacteria
Catalogus # HACH2432427
Leverancier: Hach
CAS-nummer:
Specificaties
- VPE:27
- Omschrijving:BART test for sulfate-reducing bacteria
Specificaties
Over dit artikel
BART biodetectors are excellent diagnostic tools to help identify the presence and activity of various bacteria. Sulfate reducing bacteria (SRB) are one of the most important agents in biofouling and can cause corrosion, clogging, fouling of the water and increased hygiene risks.
- Simple yet effective method for monitoring the population size and/or activity of specific groups of bacteria
- Easy to use, requiring no elaborate or costly equipment and no specialised training
- Effective and affordable tests are easy to interpret and can be performed at room temperature in virtually any environment