ContactUsLinkComponent
Micro-splitter valves, Upchurch Scientific®
  UPCHP-450
undefined
Micro-splitter valves, Upchurch Scientific®
  UPCHP-450
  • Omschrijving:
    P-450 micro-splitter valve, medium pressure, 1/4-28 threads, biocompatible, 2,1 / 4,1 µl (closed / fully open) internal volume
  • VPE:
    1
Designed to accurately split and control the low flow stream of a single incoming supply.
  • For interfacing LC-MS systems
  • Pressure rated to 800 psi (55 bar)
  • Adjustable split stream flow rates