Infusion filter, Intrapur® Plus
Infusion filters, 0,2 µm
Catalogus # BRAU4183916
Leverancier: B. Braun
Ga naar:
Product Details & Documents
Intrapur plus offers reliable air venting regardless of position with connection lines at both sides of the filter with Luer lock fittings.
- High throughput
- PVC free
- Positively charged membrane
Caution: Should not be used for more than 96 hours.
Specificaties
- Membraan:0,2 µm
- Omschrijving:Infusion filters