ContactUsLinkComponent
Carboys, with tubulation, pinchcock and handles, HDPE
  SEMA1027
 :  SEMADENI
undefined
Carboys, with tubulation, pinchcock and handles, HDPE
  SEMA1027
 :  SEMADENI
  • Ø uitw.:
    165 mm
  • Capaciteit:
    5 l
  • Hoogte:
    340 mm
  • Hals-Ø:
    50 mm
  • VPE:
    1
HDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
With silicone tubing and stainless steel pinchcock.