Organische referentiestandaard, 4,4'-Dichlorobenzophenone 10 µg/ml in cyclohexane
Catalogus # EHERL12410000CY
Leverancier: EHRENSTORFER
CAS-nummer:
Specificaties
- Verpakking:Glass bottle
- VPE:10 mL
- CAS No.:90-98-2
- Omschrijving:4,4'-Dichlorobenzophenone 10 µg/ml in cyclohexane
- Formule:C₁₃H₈Cl₂O
- Omschrijving:4,4'-Dichlorobenzophenone