ContactUsLinkComponent
Measuring beakers with handle, coloured
Measuring beakers with handle, coloured
Catalog # 213-3749
Supplier:  VITLAB
Supplier #: 481942
undefined
Measuring beakers with handle, coloured
Catalog # 213-3749
Supplier:  VITLAB
Supplier #: 481942
Log in for availability
Select an Option
100 mm
Select an Option
10 ml
Select an Option
500 ml
Select an Option
blauw
Select an Option
Select an Option
140 mm
Select an Option
01
Select an Option
12
About this item
PP, transparent.
  • Stackable
  • Cleaning up to max. 60 °C is recommended to preserve marks and inscriptions
  • Available in different colours
Ideal for food stuffs. With readable printed scale.
Frequently Bought Together