Mailing container with screw cap
Mailing containers, helder, PP/PE
Catalogus # 630-2769
Leverancier: MARIENFELD
Ga naar:
Product Details & Documents
Clear polypropylene container with polyethylene cap for use in pathology (20 ml).
Ø×H: 31×48 mm
Specificaties
- Kleur:helder
- Omschrijving:Mailing containers
- Materiaal:PP/PE